2-(3-Fluorophenyl)azepane

Supplier: Matrix Scientific

040759-500MG
101850-120EA 193.85 USD
101850-120
2-(3-Fluorophenyl)azepane
2-(3-Fluorophenyl)azepane

Hazard: Irritant

Formula: C₁₂H₁₆FN
MDL Number: MFCD02663778
CAS Number: 383130-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR