1-(1-Adamantyl)-1H-pyrazol-4-amine

Supplier: Matrix Scientific

040758-500MG
101850-118EA 484.62 USD
101850-118
1-(1-Adamantyl)-1H-pyrazol-4-amine
1-(1-Adamantyl)-1H-pyrazol-4-amine

Hazard: Irritant

Formula: C₁₃H₁₉N₃
MDL Number: MFCD12028402
CAS Number: 1172870-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR