4-Amino-N-1-dimethyl-1H-pyrazole-3-carboxamide

Supplier: Matrix Scientific

040755-500MG
101850-112EA 538.46 USD
101850-112
4-Amino-N-1-dimethyl-1H-pyrazole-3-carboxamide
4-Amino-N-1-dimethyl-1H-pyrazole-3-carboxamide

Hazard: Irritant

Formula: C₆H₁₀N₄O
MDL Number: MFCD04969452
CAS Number: 1001500-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR