2-(4-Amino-1H-pyrazol-1-yl)acetohydrazide hydrochloride

Supplier: Matrix Scientific

040754-500MG
101850-110EA 484.62 USD
101850-110
2-(4-Amino-1H-pyrazol-1-yl)acetohydrazide hydrochloride
2-(4-Amino-1H-pyrazol-1-yl)acetohydrazide hydrochloride

Hazard: Irritant

Formula: C₅H₁₀ClN₅O
MDL Number: MFCD17676097
CAS Number: 1256264-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR