[(5-Isobutylisoxazol-3-yl)methyl]amine

Supplier: Matrix Scientific

040753-500MG
101850-108EA 193.85 USD
101850-108
[(5-Isobutylisoxazol-3-yl)methyl]amine
[(5-Isobutylisoxazol-3-yl)methyl]amine

Hazard: Irritant

Formula: C₈H₁₄N₂O
MDL Number: MFCD08701261
CAS Number: 893639-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR