5-Methyl-3-pyrrolidin-2-ylisoxazole

Supplier: Matrix Scientific

040752-500MG
101850-106EA 484.62 USD
101850-106
5-Methyl-3-pyrrolidin-2-ylisoxazole
5-Methyl-3-pyrrolidin-2-ylisoxazole

Hazard: Irritant

Formula: C₈H₁₂N₂O
MDL Number: MFCD09863362
CAS Number: 1000932-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR