(3-Methyl-4,5-dihydroisoxazol-5-yl)methanol

Supplier: Matrix Scientific

040751-500MG
101850-104EA 484.62 USD
101850-104
(3-Methyl-4,5-dihydroisoxazol-5-yl)methanol
(3-Methyl-4,5-dihydroisoxazol-5-yl)methanol

Hazard: Irritant

Formula: C₅H₉NO₂
MDL Number: MFCD01612656
CAS Number: 77790-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR