(3-Isopropyl-4,5-dihydroisoxazol-5-yl)methanol

Supplier: Matrix Scientific

040750-500MG
101850-102EA 375.38 USD
101850-102
(3-Isopropyl-4,5-dihydroisoxazol-5-yl)methanol
(3-Isopropyl-4,5-dihydroisoxazol-5-yl)methanol

Hazard: Irritant

Formula: C₇H₁₃NO₂
MDL Number: MFCD12028399
CAS Number: 1142210-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR