[(5-Ethylisoxazol-3-yl)methyl]amine hydrochloride

Supplier: Matrix Scientific

040748-500MG
101850-098EA 484.62 USD
101850-098
[(5-Ethylisoxazol-3-yl)methyl]amine hydrochloride
[(5-Ethylisoxazol-3-yl)methyl]amine hydrochloride

Hazard: Irritant

Formula: C₆H₁₁ClN₂O
MDL Number: MFCD22041639
CAS Number: 1374659-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR