[(3-Ethylisoxazol-5-yl)methyl]methylamine

Supplier: Matrix Scientific

040747-500MG
101850-096EA 290.77 USD
101850-096
[(3-Ethylisoxazol-5-yl)methyl]methylamine
[(3-Ethylisoxazol-5-yl)methyl]methylamine

Hazard: Irritant

Formula: C₇H₁₂N₂O
MDL Number: MFCD11054022
CAS Number: 942519-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR