3-Isopropyl-5-pyrrolidin-2-ylisoxazole

Supplier: Matrix Scientific

040744-500MG
101850-090EA 484.62 USD
101850-090
3-Isopropyl-5-pyrrolidin-2-ylisoxazole
3-Isopropyl-5-pyrrolidin-2-ylisoxazole

Hazard: Irritant

Formula: C₁₀H₁₆N₂O
MDL Number: MFCD09701954
CAS Number: 1018163-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR