1-Benzyl-5-methyl-1H-pyrazol-3-amine

Supplier: Matrix Scientific

040737-500MG
101850-076EA 338.46 USD
101850-076
1-Benzyl-5-methyl-1H-pyrazol-3-amine
1-Benzyl-5-methyl-1H-pyrazol-3-amine

Hazard: Irritant

Formula: C₁₁H₁₃N₃
MDL Number: MFCD04969984
CAS Number: 956729-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR