N-[(1,3-Dimethyl-1H-pyrazol-4-yl)methyl]cyclopropanamine

Supplier: Matrix Scientific

040736-500MG
101850-074EA 484.62 USD
101850-074
N-[(1,3-Dimethyl-1H-pyrazol-4-yl)methyl]cyclopropanamine
N-[(1,3-Dimethyl-1H-pyrazol-4-yl)methyl]cyclopropanamine

Hazard: Irritant

Formula: C₉H₁₅N₃
MW: 165.24 g/mol
MDL Number: MFCD09701801
CAS Number: 1170202-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR