(5-Isopropylisoxazol-3-yl)methanol

Supplier: Matrix Scientific

040734-500MG
101850-070EA 290.77 USD
101850-070
(5-Isopropylisoxazol-3-yl)methanol
(5-Isopropylisoxazol-3-yl)methanol

Hazard: Irritant

Formula: C₇H₁₁NO₂
MDL Number: MFCD09859255
CAS Number: 123770-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR