[5-(4-Methoxyphenyl)-3-isoxazolyl]methanol

Supplier: Matrix Scientific

040733-500MG
101850-068EA 484.62 USD
101850-068
[5-(4-Methoxyphenyl)-3-isoxazolyl]methanol
[5-(4-Methoxyphenyl)-3-isoxazolyl]methanol

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MDL Number: MFCD06199341
CAS Number: 58492-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR