[5-(4-Methylphenyl)isoxazol-3-yl]methanol

Supplier: Matrix Scientific

040732-500MG
101850-066EA 484.62 USD
101850-066
[5-(4-Methylphenyl)isoxazol-3-yl]methanol
[5-(4-Methylphenyl)isoxazol-3-yl]methanol

Hazard: Irritant

Formula: C₁₁H₁₁NO₂
MDL Number: MFCD06199339
CAS Number: 640291-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR