5-(4-Chlorophenyl)isoxazole-3-carbohydrazide

Supplier: Matrix Scientific

040721-500MG
101850-044EA 484.62 USD
101850-044
5-(4-Chlorophenyl)isoxazole-3-carbohydrazide
5-(4-Chlorophenyl)isoxazole-3-carbohydrazide

Hazard: Irritant

Formula: C₁₀H₈ClN₃O₂
MDL Number: MFCD08446411
CAS Number: 91587-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR