5-(2,4-Dichlorophenyl)-3-isoxazolecarboxylic acid

Supplier: Matrix Scientific

040719-500MG
101850-040EA 290.77 USD
101850-040
5-(2,4-Dichlorophenyl)-3-isoxazolecarboxylic acid
5-(2,4-Dichlorophenyl)-3-isoxazolecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₅Cl₂NO₃
CAS Number: 712348-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR