5-(4-Chlorophenyl)isoxazole-3-carboxylic acid

Supplier: Matrix Scientific

040715-500MG
101850-032EA 387.69 USD
101850-032
5-(4-Chlorophenyl)isoxazole-3-carboxylic acid
5-(4-Chlorophenyl)isoxazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆ClNO₃
MDL Number: MFCD06010034
CAS Number: 33282-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR