Methyl-5-(4-hydroxyphenyl)isoxazole-3-carboxylate

Supplier: Matrix Scientific

040714-500MG
101850-030EA 444.62 USD
101850-030
Methyl-5-(4-hydroxyphenyl)isoxazole-3-carboxylate
Methyl-5-(4-hydroxyphenyl)isoxazole-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₉NO₄
MDL Number: MFCD08106871
CAS Number: 60640-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR