4-(5-Methyl-3-nitro-1H-pyrazol-1-yl)butanoic acid

Supplier: Matrix Scientific

040713-500MG
101850-028EA 484.62 USD
101850-028
4-(5-Methyl-3-nitro-1H-pyrazol-1-yl)butanoic acid
4-(5-Methyl-3-nitro-1H-pyrazol-1-yl)butanoic acid

Hazard: Irritant

Formula: C₈H₁₁N₃O₄
MDL Number: MFCD03834506
CAS Number: 696646-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR