4-(4-Chloro-5-methyl-3-nitro-1H-pyrazol-1-yl)butanoic acid
Supplier: Matrix Scientific
040710-500MG
101850-022EA
387.69
USD
101850-022
4-(4-Chloro-5-methyl-3-nitro-1H-pyrazol-1-yl)butanoic acid
4-(4-Chloro-5-methyl-3-nitro-1H-pyrazol-1-yl)butanoic acid
Hazard: Irritant
Formula:
C₈H₁₀ClN₃O₄ |
MDL Number:
MFCD03768180 CAS Number: 696646-61-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...