[4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid

Supplier: Matrix Scientific

040708-500MG
101850-018EA 387.69 USD
101850-018
[4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid
[4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetic acid

Hazard: Irritant

Formula: C₇H₆BrF₃N₂O₂
MDL Number: MFCD00682551
CAS Number: 299405-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR