4-[(4-Nitro-1H-pyrazol-1-yl)methyl]benzoic acid

Supplier: Matrix Scientific

040705-500MG
101850-012EA 387.69 USD
101850-012
4-[(4-Nitro-1H-pyrazol-1-yl)methyl]benzoic acid
4-[(4-Nitro-1H-pyrazol-1-yl)methyl]benzoic acid

Hazard: Irritant

Formula: C₁₁H₉N₃O₄
MDL Number: MFCD02090843
CAS Number: 514801-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR