4-(1,3-Benzodioxol-5-yl)-6-(trifluoromethyl)pyrimidine-2-thiol

Supplier: Matrix Scientific

040697-500MG
101849-996EA 387.69 USD
101849-996
4-(1,3-Benzodioxol-5-yl)-6-(trifluoromethyl)pyrimidine-2-thiol
4-(1,3-Benzodioxol-5-yl)-6-(trifluoromethyl)pyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₂H₇F₃N₂O₂S
MDL Number: MFCD02736869
CAS Number: 505053-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR