tert-Butyl-7-bromo-2,3-dihydro-1,4-benzoxazepine-4(5H)-carboxylate

Supplier: Matrix Scientific

040693-500MG
101849-988EA 461.54 USD
101849-988
tert-Butyl-7-bromo-2,3-dihydro-1,4-benzoxazepine-4(5H)-carboxylate
tert-Butyl-7-bromo-2,3-dihydro-1,4-benzoxazepine-4(5H)-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₈BrNO₃
MW: 328.21 g/mol
MDL Number: MFCD12028392
CAS Number: 740842-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR