3-(2-Chlorophenyl)-3-[3-(methoxymethoxy)phenyl]propanoic acid

Supplier: Matrix Scientific

040690-500MG
101849-982EA 335.38 USD
101849-982
3-(2-Chlorophenyl)-3-[3-(methoxymethoxy)phenyl]propanoic acid
3-(2-Chlorophenyl)-3-[3-(methoxymethoxy)phenyl]propanoic acid

Hazard: Irritant

Formula: C₁₇H₁₇ClO₄
MDL Number: MFCD12028390
CAS Number: 1142210-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR