N-Methyl-2-[5-(2-methylphenyl)-1,2,4-oxadiazol-3-yl]ethanamine

Supplier: Matrix Scientific

040689-500MG
101849-980EA 387.69 USD
101849-980
N-Methyl-2-[5-(2-methylphenyl)-1,2,4-oxadiazol-3-yl]ethanamine
N-Methyl-2-[5-(2-methylphenyl)-1,2,4-oxadiazol-3-yl]ethanamine

Hazard: Irritant

Formula: C₁₂H₁₅N₃O
MDL Number: MFCD12028389
CAS Number: 1142210-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR