tert-Butyl-3-formyl-1-methyl-1,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate

Supplier: Matrix Scientific

040684-500MG
101849-970EA 430.77 USD
101849-970
tert-Butyl-3-formyl-1-methyl-1,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate
tert-Butyl-3-formyl-1-methyl-1,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₉N₃O₃
MW: 265.31 g/mol
MDL Number: MFCD12028384
CAS Number: 1142210-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR