5-tert-Butyl-3-ethyl-1-methyl-1,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-3,5-dicarboxylate

Supplier: Matrix Scientific

Synonyms: 5-tert-Butyl 3-ethyl 1-methyl-6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-3,5(4H)-dicarboxylate

040683-500MG
101849-968EA 387.69 USD
101849-968
5-tert-Butyl-3-ethyl-1-methyl-1,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-3,5-dicarboxylate
5-tert-Butyl-3-ethyl-1-methyl-1,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-3,5-dicarboxylate

Hazard: Irritant

Formula: C₁₅H₂₃N₃O₄
MW: 309.37 g/mol
MDL Number: MFCD12028383
CAS Number: 1142210-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR