3-(1,3-Benzodioxol-5-yl)-3-phenylpropanoic acid

Supplier: Matrix Scientific

040680-500MG
101849-962EA 387.69 USD
101849-962
3-(1,3-Benzodioxol-5-yl)-3-phenylpropanoic acid
3-(1,3-Benzodioxol-5-yl)-3-phenylpropanoic acid

Hazard: Irritant

Formula: C₁₆H₁₄O₄
MDL Number: MFCD02058022
CAS Number: 364339-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR