3-(3-Chlorophenyl)-3-phenylpropanoic acid

Supplier: Matrix Scientific

040679-500MG
101849-960EA 387.69 USD
101849-960
3-(3-Chlorophenyl)-3-phenylpropanoic acid
3-(3-Chlorophenyl)-3-phenylpropanoic acid

Hazard: Irritant

Formula: C₁₅H₁₃ClO₂
MDL Number: MFCD09889818
CAS Number: 21998-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR