Ethyl-3-piperidin-4-ylpropanoate

Supplier: Matrix Scientific

040676-500MG
101849-954EA 484.62 USD
101849-954
Ethyl-3-piperidin-4-ylpropanoate
Ethyl-3-piperidin-4-ylpropanoate

Hazard: Irritant

Formula: C₁₀H₁₉NO₂
MDL Number: MFCD12028379
CAS Number: 71879-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR