N-(3,4-Dimethylbenzyl)-4-methylpiperidine-4-carboxamide hydrochloride
Supplier: Matrix Scientific
040675-500MG
101849-952EA
387.69
USD
101849-952
N-(3,4-Dimethylbenzyl)-4-methylpiperidine-4-carboxamide hydrochloride
N-(3,4-Dimethylbenzyl)-4-methylpiperidine-4-carboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₅ClN₂O |
MDL Number:
MFCD12028378 CAS Number: 1185302-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...