3-(4-Chlorophenyl)-3-phenylpropanoic acid

Supplier: Matrix Scientific

040673-500MG
101849-948EA 387.69 USD
101849-948
3-(4-Chlorophenyl)-3-phenylpropanoic acid
3-(4-Chlorophenyl)-3-phenylpropanoic acid

Hazard: Irritant

Formula: C₁₅H₁₃ClO₂
MDL Number: MFCD09889759
CAS Number: 21998-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR