3-[3-(2,4-Dimethoxyphenyl)-1-phenyl-1H-pyrazol-4-yl]propanoic acid

Supplier: Matrix Scientific

040667-500MG
101849-936EA 290.77 USD
101849-936
3-[3-(2,4-Dimethoxyphenyl)-1-phenyl-1H-pyrazol-4-yl]propanoic acid
3-[3-(2,4-Dimethoxyphenyl)-1-phenyl-1H-pyrazol-4-yl]propanoic acid

Hazard: Irritant

Formula: C₂₀H₂₀N₂O₄
MDL Number: MFCD12028373
CAS Number: 1170524-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR