3-[3-(1,3-Benzodioxol-5-yl)-1-phenyl-1H-pyrazol-4-yl]propanoic acid

Supplier: Matrix Scientific

040666-500MG
101849-934EA 290.77 USD
101849-934
3-[3-(1,3-Benzodioxol-5-yl)-1-phenyl-1H-pyrazol-4-yl]propanoic acid
3-[3-(1,3-Benzodioxol-5-yl)-1-phenyl-1H-pyrazol-4-yl]propanoic acid

Hazard: Irritant

Formula: C₁₉H₁₆N₂O₄
MDL Number: MFCD12028372
CAS Number: 1171384-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR