N-Methyl-N-(tetrahydrofuran-2-yl)piperidin-4-amine

Supplier: Matrix Scientific

040655-500MG
101849-918EA 484.62 USD
101849-918
N-Methyl-N-(tetrahydrofuran-2-yl)piperidin-4-amine
N-Methyl-N-(tetrahydrofuran-2-yl)piperidin-4-amine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O
MDL Number: MFCD12028361
CAS Number: 1142210-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR