N-Methyl-N-(2-morpholin-4-ylethyl)piperidin-4-amine

Supplier: Matrix Scientific

040654-500MG
101849-916EA 484.62 USD
101849-916
N-Methyl-N-(2-morpholin-4-ylethyl)piperidin-4-amine
N-Methyl-N-(2-morpholin-4-ylethyl)piperidin-4-amine

Hazard: Irritant

Formula: C₁₂H₂₅N₃O
MDL Number: MFCD12028360
CAS Number: 1142210-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR