N-Methyl-2-[5-(4-methylphenyl)-1,2,4-oxadiazol-3-yl]ethanamine hydrochloride

Supplier: Matrix Scientific

040640-500MG
101849-892EA 484.62 USD
101849-892
N-Methyl-2-[5-(4-methylphenyl)-1,2,4-oxadiazol-3-yl]ethanamine hydrochloride
N-Methyl-2-[5-(4-methylphenyl)-1,2,4-oxadiazol-3-yl]ethanamine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClN₃O
MDL Number: MFCD12028346
CAS Number: 1185296-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR