N-Phenyl-N-(2,3,4,5-tetrahydro-1,4-benzoxazepin-7-ylmethyl)cyclopropanecarboxamide hydrochloride

Supplier: Matrix Scientific

040637-500MG
101849-888EA 290.77 USD
101849-888
N-Phenyl-N-(2,3,4,5-tetrahydro-1,4-benzoxazepin-7-ylmethyl)cyclopropanecarboxamide hydrochloride
N-Phenyl-N-(2,3,4,5-tetrahydro-1,4-benzoxazepin-7-ylmethyl)cyclopropanecarboxamide hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₃ClN₂O₂
MDL Number: MFCD12028343
CAS Number: 1185302-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR