N-Methyl-2-(5-phenyl-1,2,4-oxadiazol-3-yl)ethanamine hydrochloride

Supplier: Matrix Scientific

040630-500MG
101849-876EA 484.62 USD
101849-876
N-Methyl-2-(5-phenyl-1,2,4-oxadiazol-3-yl)ethanamine hydrochloride
N-Methyl-2-(5-phenyl-1,2,4-oxadiazol-3-yl)ethanamine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄ClN₃O
MW: 239.71 g/mol
MDL Number: MFCD12028336
CAS Number: 71088-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR