1-Methyl-3-(pyrrolidin-1-ylcarbonyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine hydrochloride

Supplier: Matrix Scientific

040629-500MG
101849-874EA 484.62 USD
101849-874
1-Methyl-3-(pyrrolidin-1-ylcarbonyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine hydrochloride
1-Methyl-3-(pyrrolidin-1-ylcarbonyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉ClN₄O
MDL Number: MFCD12028335
CAS Number: 1185302-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR