4-(2-Aminoethyl)-N-(3-methylphenyl)piperidine-1-carboxamide hydrochloride

Supplier: Matrix Scientific

040616-500MG
101849-856EA 387.69 USD
101849-856
4-(2-Aminoethyl)-N-(3-methylphenyl)piperidine-1-carboxamide hydrochloride
4-(2-Aminoethyl)-N-(3-methylphenyl)piperidine-1-carboxamide hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClN₃O
MDL Number: MFCD12028322
CAS Number: 1185300-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR