3-[(2-Chloro-4-fluorobenzyl)oxy]azetidine hydrochloride

Supplier: Matrix Scientific

040615-500MG
101849-854EA 484.62 USD
101849-854
3-[(2-Chloro-4-fluorobenzyl)oxy]azetidine hydrochloride
3-[(2-Chloro-4-fluorobenzyl)oxy]azetidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂Cl₂FNO
MDL Number: MFCD12028321
CAS Number: 1185298-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR