3-[(4-Chlorobenzyl)oxy]azetidine hydrochloride

Supplier: Matrix Scientific

040611-500MG
101849-848EA 404.62 USD
101849-848
3-[(4-Chlorobenzyl)oxy]azetidine hydrochloride
3-[(4-Chlorobenzyl)oxy]azetidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃Cl₂NO
MDL Number: MFCD11849077
CAS Number: 897019-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR