3-(Benzyloxy)azetidine benzenesulfonic acid salt

Supplier: Matrix Scientific

040607-500MG
101849-842EA 484.62 USD
101849-842
3-(Benzyloxy)azetidine benzenesulfonic acid salt
3-(Benzyloxy)azetidine benzenesulphonic acid salt

Hazard: Irritant

Formula: C₁₆H₁₉NO₄S
MDL Number: MFCD11041257
CAS Number: 897019-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR