[2-(4-Methylphenyl)-4-oxo-2,4-dihydro-5H-pyrazolo[3,4-d]pyrimidin-5-yl]acetic acid

Supplier: Matrix Scientific

040598-500MG
101849-826EA 387.69 USD
101849-826
[2-(4-Methylphenyl)-4-oxo-2,4-dihydro-5H-pyrazolo[3,4-d]pyrimidin-5-yl]acetic acid
[2-(4-Methylphenyl)-4-oxo-2,4-dihydro-5H-pyrazolo[3,4-d]pyrimidin-5-yl]acetic acid

Hazard: Irritant

Formula: C₁₄H₁₂N₄O₃
MDL Number: MFCD12028306
CAS Number: 1142210-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR