3-[1-(4-Methoxyphenyl)-3-methyl-1H-1,2,4-triazol-5-yl]propanoic acid

Supplier: Matrix Scientific

040580-500MG
101849-792EA 484.62 USD
101849-792
3-[1-(4-Methoxyphenyl)-3-methyl-1H-1,2,4-triazol-5-yl]propanoic acid
3-[1-(4-Methoxyphenyl)-3-methyl-1H-1,2,4-triazol-5-yl]propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₅N₃O₃
MDL Number: MFCD12028288
CAS Number: 1142210-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR