3-[5-(Anilinocarbonyl)-1,3,4-thiadiazol-2-yl]propanoic acid

Supplier: Matrix Scientific

040548-500MG
101849-730EA 387.69 USD
101849-730
3-[5-(Anilinocarbonyl)-1,3,4-thiadiazol-2-yl]propanoic acid
3-[5-(Anilinocarbonyl)-1,3,4-thiadiazol-2-yl]propanoic acid

Hazard: Irritant

Formula: C₁₂H₁₁N₃O₃S
MDL Number: MFCD12028256
CAS Number: 1142209-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR